Learn More
Troemner™ Slotted Stainless-Steel Individual Weights
SCI_ED
Designed to stack on weight hanger.
Supplier:  Troemner™ 30390751
 View more versions of this product
Catalog No. S40390A
Add to Cart
Description
Description
  • Adjusted to tolerance specified by ANSI™/ASTM™ Class 7
Order Info
Weight hanger available separately (Cat. No. S97603A).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Metric Slotted Weight
Stainless Steel
7.85g/cm3 at 20°C
300 mg
4.11 cm
ANSI/ASTM Class 7
500 g
500 g
1.62 in.
Each
Product Suggestions
Product Suggestions