Learn More
Troemner™ Slotted Stainless-Steel Individual Weights
Designed to stack on weight hanger.
Supplier: Troemner™ 30390755
Description
- Adjusted to tolerance specified by ANSI™/ASTM™ Class 7
Order Info
Weight hanger available separately (Cat. No. S97603A).
Specifications
Metric Slotted Weight | |
Stainless Steel | |
7.85g/cm3 at 20°C | |
33 mg | |
4.11 cm |
ANSI/ASTM Class 7 | |
20 g | |
20 g | |
1.62 in. | |
Each |