Learn More
Hardy Diagnostics™ TRIO.BAS™ Air Sampler Kit
![SDP](/content/dam/fishersci/glyphs/sdp.png)
The TRIO.BAS™ instruments utilize the impaction method of air sampling. The air passing through an aspirating head, impacts onto the appropriate agar plate that collects organic substances present in sampled area.
$5972.00 - $7153.75
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
23-001-774
![]() |
Hardy Diagnostics™
BAS163K |
Charging Cable |
Each for $5,972.00
|
|
|||||
23-001-780
![]() |
Hardy Diagnostics™
BAS206K |
Induction Base |
Each for $7,153.75
|
|
|||||
Description
- All of our air samplers are lightweight, with an antibacterial techno-polymer shockproof body, have a long-lasting battery, and are IP65 protected from dust and water.
- Includes battery charger
- Protection stainless steel cover head for the aspirating head; autoclavable.
- Carrying case to protect the sampler in transit
Specifications
Air Sampler | |
200 L/min. |