Learn More
Hardy Diagnostics™ TRIO.BAS™ Air Sampler Kit
![SDP](/content/dam/fishersci/glyphs/sdp.png)
The TRIO.BAS™ instruments utilize the impaction method of air sampling. The air passing through an aspirating head, impacts onto the appropriate agar plate that collects organic substances present in sampled area.
Supplier: Hardy Diagnostics™ BAS206K
This item is not returnable.
View return policy
Description
- All of our air samplers are lightweight, with an antibacterial techno-polymer shockproof body, have a long-lasting battery, and are IP65 protected from dust and water.
- Includes battery charger
- Protection stainless steel cover head for the aspirating head; autoclavable.
- Carrying case to protect the sampler in transit
Specifications
Induction Base | |
Air Sampler | |
200 L/min. |