Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diversified Spaces™ Trapezoid Science Table
With steel frame supporting the apron.
Supplier: Diversified Spaces™ TZ7602K36C
Description
- Sturdy construction with 2.25 in. solid legs
- Steel frame supporting apron
- Chemical-resistant, non-emitting UV finish
Specifications
Trapezoid Science Table | |
24 x 60 x 36 in. | |
Steel |
Solid oak apron | |
Chemical-resistant, non-emitting UV | |
ChemGuard |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction