Learn More
Tosoh Bioscience AIA-PACK™ Test Cups - T4 (Thyroxine)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
$687.00
Specifications
For Use With (Application) | Test cup technology provides consistent and accurate assay results since all assays run the same way, every time |
---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
21-279-215
|
Tosoh Bioscience
025258 |
5 trays of 20 cups | T4 Test Cup |
Each for $687.00
|
|
21-279-49
|
Tosoh Bioscience
020358 |
12 x 1 mL | T4 Calibrator Set |
Each for $372.00
|
N/A |
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
21-279-87
|
Tosoh Bioscience
020558 |
5 trays of 20 cups | Sample Diluting Solution |
Each for $164.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on Tosoh AIA™ systems
- Saves hands-on time for operators and reduces waste
- No pre-mixing or on-board refrigeration
- Calibration stability lasts up to 90 days on most assays
Specifications
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time |