Learn More
Tosoh Bioscience AIA-PACK™ Test Cups - T4 (Thyroxine)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
Supplier: Tosoh Bioscience 025258
Description
- These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on Tosoh AIA™ systems
- Saves hands-on time for operators and reduces waste
- No pre-mixing or on-board refrigeration
- Calibration stability lasts up to 90 days on most assays
Specifications
T4 Test Cup | |
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time | |
5 trays of 20 cups |