Learn More
Tosoh Bioscience AIA-PACK™ Test Cups - PSA (Prostate Specific Antigen)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
Supplier:  Tosoh Bioscience 020663
 View more versions of this product
Catalog No. 21-279-74
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
  • These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on TOSOH AIA systems
  • Saves hands-on time for operators and reduces waste
  • No pre-mixing or on-board refrigeration
  • Calibration stability lasts up to 90 days on most assays
Specifications
Specifications
PSA Calibration Verification/Linearity Test Set
Prostate cancer
Each