Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tosoh Bioscience AIA-PACK™ Test Cups - FER (Ferritin)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
$145.00
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
21-279-30
|
Tosoh Bioscience
020353 |
1 mL | Ferritin Calibrator Set |
Each for $157.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
21-279-56
|
Tosoh Bioscience
020553 |
4 mL | Ferritin Sample Diluting Solution |
Each for $145.00
|
|
||||
21-279-73
|
Tosoh Bioscience
020653 |
Each | Ferritin Calibration Verification/Linearity Test Set |
Each for $145.00
|
|
||||
21-280-020
|
Tosoh Bioscience
025253 |
Each | Ferritin Assay Test Cup |
Each for $1,381.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on Tosoh AIA™ systems
- Saves hands-on time for operators and reduces waste
- No pre-mixing or on-board refrigeration
- Calibration stability lasts up to 90 days on most assays
Safety and Handling
ShelfLife : 1 Year
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title