Learn More
MilliporeSigma™ Supelco™ TLC Plate Rack
Holds two 20 x 20 cm, 10 x 20 cm, or 5 x 20 cm TLC plates in a standard glass developing chamber.
Supplier: MilliporeSigma™ Supelco™ Z2660271EA
Specifications
Stainless Steel | |
TLC | |
Two 20 x 20 cm, 10 x 20 cm, or 5 x 20 cm TLC plates in a standard glass developing chamber |