Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ RadEye™ Chargers

Reduce the risk of running out of battery power during critical missions with Thermo Scientific™ RadEye™ Chargers.

Supplier:  Thermo Scientific™ 425069130

 View more versions of this product

Catalog No. 17-000-846


Add to Cart

Description

Description

Make sure your portable radiation detector is always ready for use with Thermo Scientific™ RadEye™ Chargers. RadEye Chargers reduce the risk of running out of battery power when you need it most for uninterrupted radiation detection and measurement. RadEye Charger models can be used in a vehicle or on a desktop, and are compatible with most RadEye radiation detectors.

  • Rugged, easy-to-use design holds up to field use requirements
  • Reduces maintenance budget Different versions to match your charging requirements
  • Compatible with off-the-shelf cables and adaptors
  • Works with most RadEye models
  • Eliminates risk of depleted batteries during critical missions
  • Uses Commercial NiMH AAA size batteries- no transport issues or high replacement costs
  • Compatible with 4th generation RadEye Bluetooth battery back
Single Unit desktop charger
  • Standard USB allows charging from AC adapter or directly from PC Charge batteries while simultaneously managing data via USB to PC/Tablet Multi Unit desktop charger
  • Charge up to 8 RadEye units from one AC outlet in less than 5 hours
  • Status indicators for each individual RadEye being charged Ideal for large installations near large radio charger bays where individual cables and plugs is difficult to manage
Single Unit vehicle charger
  • Cradle and rubber strap designed to hold RadEye and withstand shocks to 30g
  • Standard USB allows charging from 12V lighter adapter or directly from PC
  • Supports horizontal, vertical, or gooseneck mounting (gooseneck hardware not included)
  • Ideal to convert RadEye into a light mobile detection system with data connections to mobile device or vehicle mounted PC/Tablet
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RadEye Desktop Multi Unit Charger
>850 mAh AAA 1.2 V NiMH; not included
5 hours
RadEye Bluetooth Low Energy adapter (425067095) also enables charging, but not IR/USB communications
One RadEye adapter included which also enables IR/USB communications to RadEye PC software
414 x 176 x 111
Charger
110/220 V AC to DC adapter
All RadEye models
X-Series not connected to probes
-25°C to 60°C (storage)
0°C to 35°C
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit