Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phusion Green High-Fidelity DNA Polymerase (2 U/µL)

The most accurate thermostable DNA Polymerase available offering superior performance for cloning and other applications requiring high fidelity.

$143.90 - $580.00

Specifications

Content And Storage Includes:
• Phusion DNA Polymerase, 2 U/µL
• 5X Phusion Green HF Buffer
• 5X Phusion Green GC Buffer
• DMSO
• 50 mM MgCl2

Both Phusion Green HF and Phusion Green GC Buffer provide 1.5 mM MgCl2 in the final 1X concentration.
Store at -20°C.
Format Tube
GC-Rich PCR Performance High
Polymerase Phusion High-Fidelity DNA Polymerase
Reaction Speed Fast
View More Specs

Includes:

Phusion High-Fidelity DNA Polymerase

  • Phusion DNA Polymerase 2U/μL
  • 5x Phusion HF Buffer
  • 5x Phusion GC Buffer
  • DMSO
  • 50 mM MgCl2 solution
Both Phusion HF Buffer and Phusion GC Buffer provide 1.5 mM MgCl2 in the final 1x concentration.

Phusion Green High-Fidelity DNA Polymerase

  • Phusion DNA Polymerase 2U/μL
  • 5x Phusion Green HF Buffer
  • 5x Phusion Green GC Buffer
  • DMSO
  • 50 mM MgCl2 solution
Both Phusion Green HF Buffer and Phusion Green GC Buffer provide 1.5 mM MgCl2 in the final 1x concentration.


Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
F534S
View Documents
Thermo Scientific™
F534S
100 U
Each for $143.90
Add to Cart
 
F534L
View Documents
Thermo Scientific™
F534L
500 U
Each for $580.00
Add to Cart
 
Description

Description

Incorporating a unique dsDNA–binding domain, Phusion DNA Polymerase amplifies DNA with accuracy and speed unattainable with standard PCR enzymes, even on the most difficult templates. Featuring an error rate 50–fold lower than that of Taq, and 6–fold lower than that of Pfu, Phusion DNA Polymerase is the most accurate thermostable polymerase available. This feature makes Phusion DNA Polymerases superior choice for cloning and other applications requiring high fidelity. The unique structure of the enzyme enables short protocol times, abundant yields and robust performance, even in the presence of PCR inhibitors. For the researcher, that translates to ease–of–use and convenience. In addition, Phusion DNA Polymerases also produce higher yields with lower enzyme amounts than other DNA polymerases.

Thermo Scientific Phusion DNA Polymerases set a gold standard for PCR performance. They offer a combination of characteristics that no other enzyme can match. Featuring an error rate 50-fold lower than that of Taq, and 6-fold lower than that of Pfu, Phusion DNA Polymerase is the most accurate thermostable polymerase available. This feature makes Phusion DNA Polymerases superior choice for cloning and other applications requiring high fidelity. The unique structure of the enzyme enables short protocol times, abundant yields and robust performance, even in the presence of PCR inhibitors. For the researcher, that translates to ease-of-use and convenience. In addition, Phusion DNA Polymerases also produce higher yields with lower enzyme amounts than other DNA polymerases.
New Phusion Green format is a combination of Phusion DNA Polymerase and 5X Phusion Green Buffers. The buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with excelent performance of Phusion DNA Polymerases and is compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.
Phusion High-Fidelity PCR Master Mixes are convenient 2X mixes designed to minimize the number of pipetting steps. Only template and primers need to be added by the user. Phusion High-Fidelity PCR Kit contains all the necessary reagents for PCR including control template and primers.


Highlights

  • Extreme fidelity (error rate 4.4 x 10-7 in Phusion HF Buffer)
  • Polymerase speed allows short extension times (15 to 30 s/kb)
  • Robust performance, minimal optimization needed
  • Increased product yields with minimal enzyme amounts
  • Direct loading on gel with Green Buffer
Applications
  • High performance PCR
  • High-fidelity PCR
  • Cloning
  • Sequencing (template generation)
  • High throughput
  • Difficult (GC-rich) templates
  • Template generation for sequencing
  • Long-range PCR
  • Cloning
  • Mutagenesis†
  • Microarray
Includes
Phusion High-Fidelity DNA Polymerase (F-530):
  • Phusion DNA Polymerase 2 U/μL
  • 5X Phusion HF Buffer
  • 5X Phusion GC Buffer
  • DMSO
  • 50 mM MgCl2 solution
    Both Phusion HF Buffer and Phusion GC Buffer provide 1.5 mM MgCl2 in the final 1X concentration.
Phusion Green High-Fidelity DNA Polymerase (F-534):
  • Phusion DNA Polymerase 2 U/μL
  • 5X Phusion Green HF Buffer
  • 5X Phusion Green GC Buffer
  • DMSO
  • 50 mM MgCl2
    Both Phusion Green HF and Phusion Green GC Buffer provide 1.5 mM MgCl2 in the final 1X concentration.
Phusion High-Fidelity PCR Master Mix with HF Buffer (F-531):
  • Phusion DNA Polymerase, 400 μM of each dNTP and 2X Phusion HF Buffer
  • DMSO
Phusion High-Fidelity PCR Master Mix with GC Buffer (F-532):
  • Phusion DNA Polymerase, 400 μM of each dNTP and 2X Phusion GC Buffer
  • DMSO

Master mixes provide 1.5 mM MgCl2 and 200 μM dNTP in final reaction concentration.

Phusion High-Fidelity PCR Kit (F-553):
  • Phusion DNA Polymerase 2 U/μl
  • 5X Phusion GC Buffer
  • 5X Phusion HF Buffer
  • dNTP mix (10 mM each)
  • MgCl2 solution (50 mM)
  • Control template (λ DNA)
  • 1.3 kb primers (4 μM each)
  • 10 kb primers (4 μM each)
  • DNA size standard
  • DMSO

Hazardous: No
Shelf Life: 0 Months,36 Months
Shipping Condition: Dry Ice
Storage buffer: 20 mM Tris-HCl (pH 7.4 at 25 °C), 0.1 mM EDTA, 1 mM DTT, 100 mM KCl, stabilizers, 200 μg/mL BSA and 50 % glycerol.
Storage Condition: −20°C

Specifications

Specifications

Includes:
• Phusion DNA Polymerase, 2 U/µL
• 5X Phusion Green HF Buffer
• 5X Phusion Green GC Buffer
• DMSO
• 50 mM MgCl2

Both Phusion Green HF and Phusion Green GC Buffer provide 1.5 mM MgCl2 in the final 1X concentration.
Store at -20°C.
High
Fast
High-Fidelity DNA Polymerase
2 U/µL
No
Standalone
Tube
Phusion High-Fidelity DNA Polymerase
Phusion
Cloning
52 X
Blunt
DNA
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit