Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phusion U Hot Start DNA Polymerase

Thermo Scientific Phusion U DNA Polymerase is a proofreading enzyme (25x Taq) with a proprietary mutation to read through uracils and incorporate dUTP

$235.50 - $918.00

Specifications

Content And Storage Includes:
• Phusion U Hot Start DNA Polymerase, 2 U/µL
• 5X Phusion HF Buffer
• 5X Phusion GC Buffer
• DMSO
Both Phusion HF Buffer and Phusion GC Buffer provide 1.5 mM MgCl2 in the final 1X concentration.

Store at -20°C.
Format Tube
GC-Rich PCR Performance High
Polymerase Phusion U Hot Start DNA Polymerase
Reaction Speed Fast
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
F555S
View Documents
Thermo Scientific™
F555S
100 Units
Each for $235.50
Add to Cart
 
F555L
View Documents
Thermo Scientific™
F555L
500 Units
Each for $918.00
Add to Cart
 
Description

Description

Thermo Scientific Phusion U DNA polymerase is a novel engineered high fidelity enzyme developed using fusion technology. Due to a proprietary mutation in the so called dUTP binding pocket of Phusion, Phusion U overcomes an important limitation of proofreading enzymes - it is able to incorporate dUTP and read through uracil present in DNA templates.
In addition to uracil usage possibility, Phusion U features all the superior properties of other Phusion DNA Polymerases - great accuracy, speed, ability to amplify long amplicons up to 20 kb and a high specificity with Affibody based hot start . These features make Phusion U Hot Start DNA Polymerase an excellent choice for such important applications as amplification of bisulphite-converted or damaged DNA as well as use of carryover contamination control.
Phusion U Hot Start DNA Polymerase is also available in the Master Mix and Green formats. Phusion U Hot Start PCR Master Mix is convenient 2X mix designed to minimize the number of pipetting steps. Only template and primers need to be added by the user. Phusion U Green format is a combination of Phusion U Hot Start DNA Polymerase and 5X Phusion Green Buffers. The buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with Phusion U performance and is compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.

Highlights

  • Accuracy - extremely high fidelity DNA polymerase (25x Taq)
  • Uracil-tolerance - engineered to incorporate dUTP and amplify uracil containing templates
  • Specificity - hot start for reduced nonspecific amplification and primer degradation.
  • Speed - short extension times (15-30 s/kb)
  • Direct loading of PCR products on gels
Applications
  • Amplification of bisulphite-converted DNA
  • Amplification of damaged or aged DNA
  • Carry-over contamination control
  • Uracil-excision based (USER) cloning methods
Includes:
Phusion U Hot Start DNA Polymerase (F-555)
  • Phusion U Hot Start DNA Polymerase, 2 U/μL
  • 5X Phusion HF Buffer
  • 5X Phusion GC Buffer
  • DMSO
Both Phusion HF Buffer and Phusion GC Buffer provide 1.5 mM MgCl2 in the final 1X concentration.

Phusion U Green Hot Start DNA Polymerase (F-556)
  • Phusion U Hot Start DNA Polymerase, 2 U/μL
  • 5X Phusion Green HF Buffer
  • 5X Phusion Green GC Buffer
  • DMSO
Both Phusion Green HF and Phusion Green GC Buffer provide 1.5 mM MgCl2 in the final 1X concentration.

Phusion U Hot Start PCR Master Mix
  • 2X Phusion U PCR Master Mix (Phusion U Hot Start DNA Polymerase, 2X reaction buffer, 3 mM MgCl2, 400 μM of each dATP, dTTP, dCTP, and dGTP)
  • Water, nuclease-free

Hazardous:No
Storage buffer:20 mM Tris-HCl (pH 7.4 at 25°C), 0.1 mM EDTA, 1 mM DTT, 100 mM KCl, stabilizers, and 50 % glycerol.
Storage Condition:-20°C

Specifications

Specifications

Includes:
• Phusion U Hot Start DNA Polymerase, 2 U/µL
• 5X Phusion HF Buffer
• 5X Phusion GC Buffer
• DMSO
Both Phusion HF Buffer and Phusion GC Buffer provide 1.5 mM MgCl2 in the final 1X concentration.

Store at -20°C.
High
Fast
Hot Start DNA Polymerase
25 X
Blunt
20 kb
Tube
Phusion U Hot Start DNA Polymerase
Phusion
2 U/µL
Built-In Hot Start
Standalone
DNA
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit