Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ Test Reagents
$3811.44
Specifications
For Use With (Application) | Therapeutic and abused drug testing, specific Proteins |
---|---|
Form | Liquid Ready-to-Use |
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
23-057-135
|
Ortho-Clinical Diagnostics™
6802214 |
300 Tests | Haptoglobin | N/A | N/A | ||||
This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-128
|
Ortho-Clinical Diagnostics™
6801736 |
300 Tests | Complement C4 |
Pack of 300 for $3,811.44
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-122
|
Ortho-Clinical Diagnostics™
6801737 |
300 Tests | Apolipoprotein A1 | N/A | N/A | ||||
This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-125
|
Ortho-Clinical Diagnostics™
6801988 |
300 Tests | Barbiturates |
Pack of 300 for $3,087.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-148
|
Ortho-Clinical Diagnostics™
6801712 |
300 Tests | Tobramycin |
Pack of 300 for $1,337.55
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-123
|
Ortho-Clinical Diagnostics™
6801738 |
300 Tests | Apolipoprotein B | N/A | N/A | ||||
This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-149
|
Ortho-Clinical Diagnostics™
6801767 |
300 Tests | Transferrin |
Pack of 300 for $3,811.44
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-136
|
Ortho-Clinical Diagnostics™
6801739 |
300 Tests | High Sensitivity C-Reactive Protein |
Pack of 300 for $7,087.54
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-130
|
Ortho-Clinical Diagnostics™
6801995 |
300 Tests | Cocaine Metabolite |
Pack of 300 for $3,087.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-057-133
|
Ortho-Clinical Diagnostics™
6801711 |
300 Tests | Gentamicin |
Pack of 300 for $4,367.93
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-057-137
|
Ortho-Clinical Diagnostics™
6801732 |
300 Tests | IgA | N/A | N/A | ||||
This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-150
|
Ortho-Clinical Diagnostics™
6801710 |
300 Tests | Valproic Acid |
Pack of 300 for $4,367.93
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-143
|
Ortho-Clinical Diagnostics™
6801997 |
300 Tests | Opiates |
Pack of 300 for $3,087.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-151
|
Ortho-Clinical Diagnostics™
6801709 |
300 Tests | Vancomycin |
Pack of 300 for $4,126.55
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-140
|
Ortho-Clinical Diagnostics™
6801740 |
300 Tests | Microalbumin |
Pack of 300 for $3,811.44
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-147
|
Ortho-Clinical Diagnostics™
6801994 |
300 Tests | Cannabinoids |
Pack of 300 for $3,087.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-129
|
Ortho-Clinical Diagnostics™
6801713 |
300 Tests | Caffeine |
Pack of 200 for $3,969.02
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-127
|
Ortho-Clinical Diagnostics™
6801735 |
300 Tests | Complement C3 |
Pack of 300 for $3,811.44
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-131
|
Ortho-Clinical Diagnostics™
6801728 |
600 Tests |
Pack of 600 for $3,223.50
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-142
|
Ortho-Clinical Diagnostics™
6801996 |
300 Tests | Methadone |
Pack of 300 for $3,087.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-146
|
Ortho-Clinical Diagnostics™
6801729 |
300 Tests | Rheumatoid Factor |
Pack of 300 for $3,811.44
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-120
|
Ortho-Clinical Diagnostics™
6802216 |
300 Tests | alpha-1-antitrypsin |
Pack of 300 for $3,811.44
|
|
||||
23-057-126
|
Ortho-Clinical Diagnostics™
6801989 |
300 Tests | Benzodiazepines |
Pack of 300 for $3,087.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-124
|
Ortho-Clinical Diagnostics™
6802218 |
300 Tests | Anti-Streptolysin |
Pack of 300 for $3,811.44
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-121
|
Ortho-Clinical Diagnostics™
6801991 |
300 Tests | Amphetamines |
Pack of 300 for $3,087.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-132
|
Ortho-Clinical Diagnostics™
6802001 |
300 Tests |
Pack of 300 for $3,444.05
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-138
|
Ortho-Clinical Diagnostics™
6801733 |
300 Tests | IgG |
Pack of 300 for $3,811.44
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-139
|
Ortho-Clinical Diagnostics™
6801734 |
300 Tests | IgM |
Pack of 300 for $3,811.44
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-057-144
|
Ortho-Clinical Diagnostics™
6801741 |
300 Tests | Prealbumin |
Pack of 300 for $3,811.44
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
Specifications
Therapeutic and abused drug testing, specific Proteins | |
Liquid Ready-to-Use |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title