Learn More
National Optical Teaching Laboratory Biological Advanced Microscope
The Stellar Series teaching and laboratory biological microscopes adopt a new structural design, with a smoother appearance and stable performance.
Supplier: National Optical DSTELLAR 1 PROT4
Description
- The Stellar Pro Series teaching biological microscopes adopts a new structural design and updated appearance.
- The Stellar PRO Series includes all of the features of the Stellar1 Series with the addition of an infinity corrected optical system for superb performance.
- Choose between binocular or trinocular heads with a choice of three objectives (4X, 10X, 40X) or four objectives which includes the 100X oil immersion lens.
Specifications
WF 10X/18 mm | |
LED | |
Stellar | |
Biological Microscope |
Trinocular | |
4X, 10X, 40X, 100X | |
Infinity Corrected |