Learn More
MilliporeSigma™ Supelco™ T-Type Gas Line Filters
Remove dust and small particles that could otherwise foul downstream needle valves
Supplier: MilliporeSigma™ Supelco™ 25806
Specifications
Removing Dust and Small Particles That Could Otherwise Foul Downstream Needle Valves | |
Replaceable 7 μm Sintered Frit, Nuts, and Ferrules | |
Brass | |
1/4 in. Male |