Learn More
Stainless Steel powder, -20+35 mesh, Type 316-L, Thermo Scientific Chemicals
$80.65 - $241.65
Specifications
Chemical Name or Material | Stainless Steel powder, Type 316-L |
---|---|
Avg. Mol. Wt. or Mol. Wt. Range | Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% |
MDL Number | MFCD00148499 |
Solubility Information | Insoluble in water. |
Mesh Size | −20+35 Mesh |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA4346036
|
Thermo Scientific Chemicals
04346036 |
500 g |
Each
for $80.65
Save 12% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA43460A3
|
Thermo Scientific Chemicals
043460A3 |
2 kg |
Each
for $241.65
Save 12% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel powder, Type 316-L | |
MFCD00148499 | |
−20+35 Mesh |
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% | |
Insoluble in water. | |
Powder |
Safety and Handling
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.