Learn More
Stainless Steel powder, -20+35 mesh, Type 316-L, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 043460A3
Description
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel powder, Type 316-L | |
MFCD00148499 | |
Insoluble in water. | |
Powder |
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% | |
2 kg | |
−20+35 Mesh |
Safety and Handling
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
TSCA : Yes
Recommended Storage : Ambient temperatures