Learn More
Stainless Steel gauze, 20 mesh woven from 0.102mm (0.004in) dia wire, Type 316, Thermo Scientific Chemicals
Open area: 84.6%; Width of opening: 0.046 in.
$35.65 - $303.65
Specifications
Chemical Name or Material | Stainless Steel |
---|---|
Appearance | Gauze |
Molecular Formula | Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% |
MDL Number | MFCD00148499 |
Solubility Information | Insoluble in water. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA40943FL
|
Thermo Scientific Chemicals
040943FL |
75 x 75 mm |
Each
for $35.65
Save 12% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA40943GT
|
Thermo Scientific Chemicals
040943GT |
150 x 150 mm |
Each
for $97.65
Save 13% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA40943HB
|
Thermo Scientific Chemicals
040943HB |
300 x 300 mm |
Each
for $303.65
Save 12% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel | |
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% | |
Insoluble in water. | |
Wire Cloth |
Gauze | |
MFCD00148499 | |
20 Mesh |
Safety and Handling
H317-H351: May cause an allergic skin reaction. Suspected of causing cancer.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.