Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stainless Steel gauze, 20 mesh woven from 0.102mm (0.004in) dia wire, Type 316, Thermo Scientific Chemicals

Open area: 84.6%; Width of opening: 0.046 in.

Supplier:  Thermo Scientific Chemicals 040943HB

 View more versions of this product

Catalog No. AA40943HB


Order in Bulk

Request bulk or custom formats

Add to Cart

Description

Description

Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Stainless Steel
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
300 x 300 mm
20 Mesh
Gauze
MFCD00148499
Insoluble in water.
Wire Cloth
Safety and Handling

Safety and Handling

H317-H351: May cause an allergic skin reaction. Suspected of causing cancer.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

RUO – Research Use Only