Learn More
Honeywell Burdick & Jackson™ SSPDS Accessory, Female Keyed Quick-Connect Coupling
Developed specifically for high-purity solvents. All internal surfaces are 316 stainless steel or PTFE. Pressure rated to 150 psig. Air inclusion of <0.01 ml and solvent spillage of <0.02 mL during coupling virtually eliminates moisture contamination and makes purging solvent lines unnecessary when changing containers.
$1664.00 - $2137.00
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
60-090-531
|
Honeywell Burdick & Jackson™
CAKEYC1BODY |
Color-coded for Key C1 |
Each for $2,137.00
|
|
|||||
60-090-534
|
Honeywell Burdick & Jackson™
CAKEYC4BODY |
Color-coded for Key C4 |
Each for $1,664.00
|
|
|||||
60-090-530
|
Honeywell Burdick & Jackson™
CAKEYC0BODY |
Color-coded for Key C0 |
Each for $2,054.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
- As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
- Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
Specifications
1 Ea. |