Learn More
Honeywell Burdick & Jackson™ SSPDS Accessory, Female Keyed Quick-Connect Coupling
Developed specifically for high-purity solvents. All internal surfaces are 316 stainless steel or PTFE. Pressure rated to 150 psig. Air inclusion of <0.01 ml and solvent spillage of <0.02 mL during coupling virtually eliminates moisture contamination and makes purging solvent lines unnecessary when changing containers.
Supplier: Honeywell Burdick & Jackson™ CAKEYC4BODY
Description
- Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
- As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
- Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color-coded for Key C4 | |
1 Ea. |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov