Learn More
Bel-Art™ Spinbar Octagon Teflon Magnetic Stirring Bars
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape.
$6.10 - $29.00
Specifications
Autoclavable | Yes |
---|---|
Certifications/Compliance | FDA, USP Class VI, ISO 9001:2008 |
Color | White |
Material | Alnico Magnet |
Shape | Octagon |
Catalog Number | Mfr. No. | Diameter (English) | Diameter (Metric) | Length (Metric) | Length (English) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) | Diameter (Metric) | Length (Metric) | Length (English) | Price | Quantity & Availability | ||
S71736
|
Bel-Art™
F371100002 |
0.31 in. | 8 mm | 50.8 mm | 2 in. |
Each for $9.70
|
|
||
S43324
|
Bel-Art™
F371100003/EMD |
0.5 in. | 12.7 mm | 76.2 mm | 3 in. |
Each for $29.00
|
|
||
S42991
|
Bel-Art™
F371100078/EMD |
0.31 in. | 8 mm | 22 mm | 0.87 in. |
Each for $7.35
|
|
||
S42992
|
Bel-Art™
F371100118/EMD |
0.31 in. | 8 mm | 28.6 mm | 1.12 in. |
Each for $6.90
|
|
||
S42993
|
Bel-Art™
F371100138/EMD |
0.37 in. | 9.5 mm | 25.4 mm | 1 in. |
Each for $10.25
|
|
||
S42994
|
Bel-Art™
F371100158/EMD |
0.31 in. | 8 mm | 41.14 mm | 1.62 in. |
Each for $10.25
|
|
||
S43325
|
Bel-Art™
F371100178/EMD |
0.37 in. | 9.5 mm | 47.5 mm | 1.87 in. |
Each for $13.50
|
|
||
S35825
|
Bel-Art™
F371100212/EMD |
0.31 in. | 8 mm | 63.5 mm | 2.5 in. |
Each for $25.50
|
|
||
S71737
|
Bel-Art™
F371101122 |
0.5 in. | 12.7 mm | 38 mm | 1.5 in. |
Each for $25.25
|
|
||
S35820
|
Bel-Art™
F371101128/EMD |
0.37 in. | 9.5 mm | 38 mm | 1.5 in. |
Each for $18.75
|
|
||
S71738
|
Bel-Art™
F371101218 |
0.11 in. | 3 mm | 12.7 mm | 0.5 in. |
Each for $6.10
|
|
||
S42995
|
Bel-Art™
F371101238/EMD |
0.37 in. | 9.5 mm | 12.7 mm | 0.5 in. |
Each for $6.90
|
|
||
S42996
|
Bel-Art™
F371101388/EMD |
0.37 in. | 9.5 mm | 35 mm | 1.37 in. |
Each for $18.75
|
|
||
S43328
|
Bel-Art™
F371102128/EMD |
0.37 in. | 9.5 mm | 63.5 mm | 2.5 in. |
Each for $16.00
|
|
||
S42997
|
Bel-Art™
F371105838/EMD |
0.37 in. | 9.5 mm | 16 mm | 0.625 in. |
Each for $8.85
|
|
||
S42998
|
Bel-Art™
F371107838/EMD |
0.37 in. | 9.5 mm | 22 mm | 0.87 in. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
Description
- Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® brand PTFE coating for superior chemical compatibility
- Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
- Pivot ring aids in reducing friction and chattering (S71738 does not have a pivot ring)
- Autoclavable
- Manufactured in registered ISO 9001:2008 facilities, as verified by SGS certification
Specifications
Yes | |
White | |
Octagon |
FDA, USP Class VI, ISO 9001:2008 | |
Alnico Magnet |