Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Liquid Safety Solutions Spilfyter™ Wipe Kit Station

These free standing Wipe Kit Stations can strategically mount your Spilfyter™ wipe kits close to the work spaces and equipment that need them the most.

Supplier:  Liquid Safety Solutions 944STATION46BL

 View more versions of this product

Catalog No. 19-066-817


Add to cart
This item is not returnable. View return policy

Description

Description

  • Free standing wipe kit stand with bucket attachment
  • Set up cleaning and disinfecting stations strategically throughout your facility or campus
  • Powder-coated steel
  • Fixture only, does not include a wipe kit
Specifications

Specifications

Wipe Kit Station
Bucket mount header: 15 x 10 in.
Fixture only, does not include a wipe kit
Black
Stand: 46 in., Bucket: 36 in.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit