Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Standard Stem Funnels
SCI_ED

Feature an exclusive filter-step internal design for speedier filtrations

$72.00 - $83.00

Specifications

Material Polypropylene
Autoclavable Yes
Length (Metric) Stem 6.5cm
View More Specs

Products 2
Catalog Number Mfr. No. Length (Metric) Stem Diameter (Metric) Filter Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Stem Diameter (Metric) Filter Capacity (Metric) Price Quantity & Availability  
S43512
SCI_ED
View Documents
Bel-Art™
F146760055/EMD
6.5cm 9cm 32.8 mL
Pack of 12 for $83.00
Add to cart
 
S43513
SCI_ED
View Documents
Bel-Art™
F146760065/EMD
6.5cm 11cm 59.1 mL
Pack of 12 for $72.00
Add to cart
 
Description

Description

  • External ribs prevent airlock
  • The parallel stem measures 8mm O.D.
  • Molded polypropylene is steam autoclavable at 121°C (250°F)

Specifications

Specifications

Polypropylene
Yes
6.5cm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit