Learn More
Bel-Art™ SP Scienceware™ Polyethylene Utility Bags
SCI_ED
Reusable, transparent bags are well-suited for shipping or storage and are especially useful as sample bags
Supplier:  Bel-Art™ H131780816/EMD
 View more versions of this product
Catalog No. S42894
Explore available promotions
Add to Cart
This item is not returnable. View return policy
Description
Description
  • FDA approved material; Latex free
  • Different sizes available to best match particular packaging requirements
  • Not autoclavable
Specifications
Specifications
Polyethylene
16 x 8 in. (406 x 203mm)
406 mm
203 mm
0.102 mm
16 in.
8 in.
Product Suggestions
Product Suggestions