Learn More
Bel-Art™ SP Scienceware™ Magic Touch 2™ Ice Pans
Superior insulating properties combined with high chemical resistance
$104.00 - $216.00
Specifications
Product Type | Laboratory Ice Pan |
---|---|
Material | Urethane |
Shape | Raised |
Surface Type | Lightly textured, Non-slip |
Temperature (English) | -320°F to 212°F |
Catalog Number | Mfr. No. | Capacity (Metric) | Color | Includes | Dimensions | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Color | Includes | Dimensions | Price | Quantity & Availability | ||
S43206
![]() |
Bel-Art™
M168071103/EMD |
1 L | Cherry Red | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 7.25 x 7.25 x 4.5 in. (18.3 x 18.3 x 11.6 cm) |
Each for $107.00
|
|
S43204
![]() |
Bel-Art™
M168071101/EMD |
1 L | True Blue | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 7.25 x 7.25 x 4.5 in. (18.3 x 18.3 x 11.6 cm) |
Each for $106.00
|
|
S43213
![]() |
Bel-Art™
M168071104/EMD |
1 L | Emerald Green | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 7.25 x 7.25 x 4.5 in. (18.3 x 18.3 x 11.6 cm) |
Each for $104.00
|
|
S43214
![]() |
Bel-Art™
M168074101/EMD |
4 L | True Blue | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 12 x 9 x 8 in. (30.4 x 22.9 x 20 cm) |
Each for $150.00
|
|
S43215
![]() |
Bel-Art™
M168074102/EMD |
4 L | Black Ebony | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 12 x 9 x 8 in. (30.4 x 22.9 x 20 cm) |
Each for $150.00
|
|
S43216
![]() |
Bel-Art™
M168074103/EMD |
4 L | Cherry Red | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 12 x 9 x 8 in. (30.4 x 22.9 x 20 cm) |
Each for $150.00
|
|
S43218
![]() |
Bel-Art™
M168079101/EMD |
9 L | True Blue | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 16.5 x 12 x 8 in. (42 x 30.3 x 20 cm) |
Each for $216.00
|
|
S43217
![]() |
Bel-Art™
M168074104/EMD |
4 L | Emerald Green | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 12 x 9 x 8 in. (30.4 x 22.9 x 20 cm) |
Each for $151.00
|
|
S43220
![]() |
Bel-Art™
M168079102/EMD |
9 L | Black Ebony | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 16.5 x 12 x 8 in. (42 x 30.3 x 20 cm) |
Each for $210.00
|
|
S43221
![]() |
Bel-Art™
M168079103/EMD |
9 L | Cherry Red | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 16.5 x 12 x 8 in. (42 x 30.3 x 20 cm) |
Each for $216.00
|
|
S43224
![]() |
Bel-Art™
M168079104/EMD |
9 L | Emerald Green | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 16.5 x 12 x 8 in. (42 x 30.3 x 20 cm) |
Each for $210.00
|
|
S43226
![]() |
Bel-Art™
M168079903/EMD |
9 L | Cherry Red | Pour spout, drip-proof rims, fill line indicator, without Lid | 16.5 x 12 x 8.5 in. (42 x 30.3 x 21.7 cm) |
Each for $172.00
|
|
S43349
![]() |
Bel-Art™
M168079901/EMD |
9 L | True Blue | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 16.5 x 12 x 8.5 in. (42 x 30.3 x 21.7 cm) |
Each for $172.00
|
|
S43351
![]() |
Bel-Art™
M168079902/EMD |
9 L | Black Ebony | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 16.5 x 12 x 8.5 in. (42 x 30.3 x 21.7 cm) |
Each for $168.00
|
|
S43352
![]() |
Bel-Art™
M168079904/EMD |
9 L | Emerald Green | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 16.5 x 12 x 8.5 in. (42 x 30.3 x 21.7 cm) |
Each for $168.00
|
|
S43205
![]() |
Bel-Art™
M168071102/EMD |
1 L | Black Ebony | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 7.25 x 7.25 x 4.5 in. (18.3 x 18.3 x 11.6 cm) | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- Compatible not only with wet ice, but also with ultra-low temperature materials including dry ice, salt slurries, dry ice solvent slurries, or liquid nitrogen
- Able to withstand a wide temperature range of -196°C (-320°F) to 100°C (212°F)
- High-quality, expanded urethane laboratory ice pans are lightweight, durable, non-sweating and impervious to moisture and odors
- Lightly textured, non-slip finish and raised edge provides secure transport and a stylish look
- Feature a convenient pour spout, drip-proof rims, a fill line indicator, and a snug-fitting, keyed lid and base for easy stacking
1L pan does not include pour spout or fill line indicator
9L pan available with or without lid
Specifications
Laboratory Ice Pan | |
Raised | |
-320°F to 212°F |
Urethane | |
Lightly textured, Non-slip | |
-196°C to 100°C |