Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ Sodium Thiosulfate, 0.011N
Sodium Thiosulfate, 0.011N is a replacement reagent.
$16.25 - $118.00
Specifications
Color | Pale yellow |
---|---|
Composition | 10102-17-7, 497-19-8 |
Concentration | 0.011 N |
Description | Sodium thiosulfate, Sodium carbonate |
Format | Liquid |
Description
Available in multiple sizes:
- Gallon size for multiple tests.
- Kit size bottle.
Specifications
Pale yellow | |
0.011 N | |
Liquid |
10102-17-7, 497-19-8 | |
Sodium thiosulfate, Sodium carbonate | |
Sodium Thiosulfate |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title