Learn More
ICU Medical Pulsator™ Plus Liquid Heparin Arterial Blood Sampling Kits
Quick-filling syringes respond to arterial pressure, aspirate easily
Supplier: ICU Medical 40362
Includes: 3-cc or 5-cc Luer lock syringe and Filter-Pro™ device in polybag or Tyvek packaging.
Description
- Pulsating effect allows for precise positioning in arterial lumen
- Syringe response to arterial pressure helps ensure arterial sample
- Rapid fill minimizes patient discomfort
- 3mL arterial blood-sampling syringes are easily aspirated during low blood pressure or code situation
- Needles with different gauges and lengths
- Prefilled with liquid sodium heparin concentration of 31 IU/mL
- Specially formulated electrolyte-balanced liquid sodium heparin offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels
- Clot-free blood sampling without risking sample dilution
- Provides consistent and accurate analysis for all parameters tested
Order Info
- With or without support kit (alcohol prep pad, iodophor prep pad, gauze pad, patient label, biohazard labeled ice bag, adhesive bandage)
- Sterile Tyvek™ packaging
- Come with Filter-Pro™ air bubble removal device
Specifications
Provides consistent and accurate analysis for all parameters tested, offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels | |
Pulsator Arterial Blood Sampling Kit | |
Polybag |
3mL luer-lock syringe and Filter-Pro™ air bubble removal device, 31 IU/mL concentration of liquid sodium heparin | |
200/Cs. | |
3 mL |