Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Smiths Medical Pulsator™ Plus Liquid Heparin Arterial Blood Sampling Kits
Quick-filling syringes respond to arterial pressure, aspirate easily
$698.50
Specifications
For Use With (Application) | Provides consistent and accurate analysis for all parameters tested, offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels |
---|---|
Product Type | Pulsator Arterial Blood Sampling Kit |
Packaging | Polybag |
Volume (Metric) | 3 mL |
Includes: 3-cc or 5-cc Luer lock syringe and Filter-Pro™ device in polybag or Tyvek packaging.
Catalog Number | Mfr. No. | Quantity | Includes | Packaging | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Includes | Packaging | Price | Quantity & Availability | |||
22-024-974
|
Smiths Medical
40391 |
100/Cs. | Pulsator™ 3mL Luer lock syringe with Filter-Pro™ device, 22g x 1.5 in. and 23g x 1 in. needles, Needle-Pro device, support kit, needle stop, and polybag packaging | Polybag |
Case of 100
for $42.86
Save 92% Reg : |
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-004-198
|
Smiths Medical
4037NP1 |
100/Cs. | Pulsator™ 3mL Luer lock syringe with Filter-Pro™ device, 20g x 1.5 in. and 22g x 1 in. needles, Needle-Pro™ device, support kit, needle stop, and polybag packaging | Polybag |
Case of 100 for $615.50
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-024-972
|
Smiths Medical
40362 |
200/Cs. | 3mL luer-lock syringe and Filter-Pro™ air bubble removal device, 31 IU/mL concentration of liquid sodium heparin | Polybag |
Case of 200 for $698.50
|
|
Description
- Pulsating effect allows for precise positioning in arterial lumen
- Syringe response to arterial pressure helps ensure arterial sample
- Rapid fill minimizes patient discomfort
- 3mL arterial blood-sampling syringes are easily aspirated during low blood pressure or code situation
- Needles with different gauges and lengths
- Prefilled with liquid sodium heparin concentration of 31 IU/mL
- Specially formulated electrolyte-balanced liquid sodium heparin offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels
- Clot-free blood sampling without risking sample dilution
- Provides consistent and accurate analysis for all parameters tested
Order Info
- With or without support kit (alcohol prep pad, iodophor prep pad, gauze pad, patient label, biohazard labeled ice bag, adhesive bandage)
- Sterile Tyvek™ packaging
- Come with Filter-Pro™ air bubble removal device
Specifications
Provides consistent and accurate analysis for all parameters tested, offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels | |
Pulsator Arterial Blood Sampling Kit | |
Polybag | |
3 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title