Learn More
Smiths Medical Pro-Vent™ Plus Arterial Blood Gas Sampling 3cc/21IU Kits
Pro-Vent™ Plus 3 mL Arterial Blood Sampling syringes utilize dry lithium heparin with a low concentration of 7 I.U. per mL. The low heparin concentration allows for healthcare professionals to obtain very small blood samples and still maintain the full range of blood gas, electrolyte, and metabolite testing with reduced heparin interference and extended clotting time.
Specifications
For Use With (Application) | Allows healthcare professionals to obtain very small blood samples and still maintain the full range of blood gas, electrolyte and metabolite testing with reduced heparin interference and extended clotting time |
---|---|
Product Type | Arterial Blood Sampling Kit |
Packaging | Sterile Tyvek™ |
Volume (Metric) | 3 mL |
Sterility | Sterile |
Catalog Number | Mfr. No. | Quantity | Includes | Needle Gauge | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Includes | Needle Gauge | Price | Quantity & Availability | |||
22-024-953
|
Smiths Medical
4698LH |
200/Cs. | 3mL luer-slip syringe with Filter-Pro™ device, 23G x 1 in. preattached needle with Needle-Pro™ needle safety device and adhesive bandage, 7 IU/mL concentration of low dry lithium heparin as anticoagulant | 23 G |
Case of 200 for $636.50
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-024-954
|
Smiths Medical
4699LH |
100/Cs. | 3mL luer-slip syringe with Filter-Pro™ device, 23G x 1 in. preattached needle with Needle-Pro™ needle safety device, 22 x 1.5 in. secondary needle with Needle-Pro™ needle safety device, support kit | 22 to 23 G |
Case of 100 for $283.50
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Advanced-formula dry lithium heparin
- Offsets binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level
- Consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, lactate
- Large venting filter media facilitates quick filling and the siliconized barrel allows easy aspiration, even in code situations
- Filter-Pro™ device permits ease of removing air bubbles without accidentally expelling blood or aerosolizing the sample
- Needle-Pro™ needle protection device (sturdy, puncture-resistant material) lock in place, completely covering used needle
- Support kits contain alcohol prep pad, lodophor prep pad, gauze pad, patient label, biohazard-labeled ice bag, adhesive bandage
Order Info
Sterile Tyvek packaging except as noted.
Specifications
Allows healthcare professionals to obtain very small blood samples and still maintain the full range of blood gas, electrolyte and metabolite testing with reduced heparin interference and extended clotting time | |
Sterile Tyvek™ | |
Sterile |
Arterial Blood Sampling Kit | |
3 mL |
Safety and Handling
ShelfLife : 3 Years
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.