Learn More
Silica Standard Solution, APHA, Spectrum™ Chemical
SDP
Supplier:  Spectrum Chemical Manufacturing Corporation S135125ML
 View more versions of this product
Catalog No. 18-613-101
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Specifications
Specifications
1.000 ± 0.010 mg SiO₂/mL
CAS: 13517-24-3; 7732-18-5
Poly Bottle
125 mL
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.