Learn More
SiliCycle™ SiliaPrepX™ SPE Cartridges - WAX
SPE Cartridges packed with a polymeric weak anion exchange (WAX) phase, for extraction of strong acidic compounds.
$208.50 - $3070.00
Specifications
Adsorbent Type | Polymer |
---|---|
Carbon Load | 0.86 |
For Use With (Application) | Extraction of strongly acidic compounds, highly stable in organic solvents |
For Use With (Equipment) | Vacuum Manifolds or Automated SPE Equipment |
Particle Size | 85 μm |
Catalog Number | Mfr. No. | Bed Weight | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Bed Weight | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
01-184-607
|
SiliCycle™
SPEP002001AA |
30 mg | 1 mL | 100 Pk. |
Pack of 100 for $279.50
|
|
|||
01-184-608
|
SiliCycle™
SPEP002001AAL |
30 mg | 1 mL | 500 Pk. |
Pack of 500 for $1,456.00
|
|
|||
01-184-822
|
SiliCycle™
SPEP002001AAM |
30 mg | 1 mL | 1000 Pk. |
Pack of 1000 for $3,070.00
|
|
|||
01-184-613
|
SiliCycle™
SPEP002003AA |
30 mg | 3 mL | 50 Pk. |
Pack of 50 for $208.50
|
|
|||
01-184-614
|
SiliCycle™
SPEP002003AAJ |
30 mg | 3 mL | 200 Pk. |
Pack of 200 for $744.00
|
|
|||
01-184-609
|
SiliCycle™
SPEP002001BB |
60 mg | 1 mL | 100 Pk. |
Pack of 100 for $316.50
|
|
|||
01-184-610
|
SiliCycle™
SPEP002001BBL |
60 mg | 1 mL | 500 Pk. |
Pack of 500 for $1,498.00
|
|
|||
01-184-615
|
SiliCycle™
SPEP002003BB |
60 mg | 3 mL | 50 Pk. |
Pack of 50 for $247.00
|
|
|||
01-184-797
|
SiliCycle™
SPEP002003BBJ |
60 mg | 3 mL | 200 Pk. |
Pack of 200 for $934.50
|
|
|||
01-184-620
|
SiliCycle™
SPEP002006BB |
60 mg | 6 mL | 30 Pk. |
Pack of 30 for $223.50
|
|
|||
01-184-621
|
SiliCycle™
SPEP002006BBJ |
60 mg | 6 mL | 200 Pk. |
Pack of 200 for $1,135.00
|
|
|||
01-184-611
|
SiliCycle™
SPEP002001C |
100 mg | 1 mL | 100 Pk. |
Pack of 100 for $344.00
|
|
|||
01-184-823
|
SiliCycle™
SPEP002003C |
100 mg | 3 mL | 50 Pk. |
Pack of 50 for $263.50
|
|
|||
01-184-616
|
SiliCycle™
SPEP002003CJ |
100 mg | 3 mL | 200 Pk. |
Pack of 200 for $994.50
|
|
|||
01-184-617
|
SiliCycle™
SPEP002003CL |
100 mg | 3 mL | 500 Pk. |
Pack of 500 for $2,360.00
|
|
|||
01-184-622
|
SiliCycle™
SPEP002006C |
100 mg | 6 mL | 30 Pk. |
Pack of 30 for $279.50
|
|
|||
01-184-618
|
SiliCycle™
SPEP002003G |
200 mg | 3 mL | 50 Pk. |
Pack of 50 for $320.50
|
|
|||
01-184-623
|
SiliCycle™
SPEP002006G |
200 mg | 6 mL | 30 Pk. |
Pack of 30 for $295.00
|
|
|||
01-184-619
|
SiliCycle™
SPEP002003P |
500 mg | 3 mL | 50 Pk. |
Pack of 50 for $555.00
|
|
|||
01-184-624
|
SiliCycle™
SPEP002006P |
500 mg | 6 mL | 30 Pk. |
Pack of 30 for $398.00
|
|
|||
01-184-625
|
SiliCycle™
SPEP002012P |
500 mg | 12 mL | 20 Pk. |
Pack of 20 for $344.00
|
|
|||
01-184-825
|
SiliCycle™
SPEP002020P |
500 mg | 25 mL | 20 Pk. |
Pack of 20 for $516.50
|
|
|||
01-184-627
|
SiliCycle™
SPEP002020S |
1 g | 25 mL | 20 Pk. |
Pack of 20 for $660.00
|
|
|||
01-184-626
|
SiliCycle™
SPEP002012U |
2 g | 12 mL | 20 Pk. |
Pack of 20 for $817.50
|
|
|||
01-184-628
|
SiliCycle™
SPEP002020U |
2 g | 25 mL | 20 Pk. |
Pack of 20 for $785.50
|
|
|||
01-184-606
|
SiliCycle™
SPEP0020011A |
10 mg | 1 mL | 100 Pk. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
01-184-612
|
SiliCycle™
SPEP0020031A |
10 mg | 3 mL | 50 Pk. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
Description
- For use with vacuum manifolds and automated SPE equipments
- Tightly packed and homogeneous silica bed for reliability and increased productivity
- Tight particle size distribution and very good packing with no fines, allowing excellent separation
- High recoveries and yields
- Reproducible flow rates lot-to-lot
- Flanged polypropylene tubes
- 20 μm polyethylene frits
Specifications
Polymer | |
Extraction of strongly acidic compounds, highly stable in organic solvents | |
85 μm | |
Polystyrene-divinylbenzene WAX | |
Polypropylene (PP) tube, Polyethylene (PE) frits | |
60 Å |
0.86 | |
Vacuum Manifolds or Automated SPE Equipment | |
SiliaPrepX | |
SPE Cartridge | |
Spherical | |
800 m2/g |