Learn More
SiliCycle™ SiliaPrepX™ SPE Cartridges - DVB
SPE Cartridges packed with polystyrene-divinylbenzene copolymer (DVB), for extraction of neutral, acidic or basic compounds.
$183.00 - $2324.00
Specifications
Adsorbent Type | Polymer |
---|---|
For Use With (Application) | Extraction of drugs and metabolites from biological fluids, API from tablets and cream |
For Use With (Equipment) | Vacuum Manifolds or Automated SPE Equipment |
Particle Size | 85 μm |
Product Line | SiliaPrepX™ |
Catalog Number | Mfr. No. | Bed Weight | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Bed Weight | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
01-182-896
|
SiliCycle™
SPEP000101AA |
30 mg | 1 mL | 100 Pk. |
Pack of 100 for $259.00
|
|
01-182-897
|
SiliCycle™
SPEP000101AAL |
30 mg | 1 mL | 500 Pk. |
Pack of 500 for $1,227.00
|
|
01-182-898
|
SiliCycle™
SPEP000101AAM |
30 mg | 1 mL | 1000 Pk. |
Pack of 1000 for $2,324.00
|
|
01-182-903
|
SiliCycle™
SPEP000103AA |
30 mg | 3 mL | 50 Pk. |
Pack of 50 for $183.00
|
|
01-182-904
|
SiliCycle™
SPEP000103AAJ |
30 mg | 3 mL | 200 Pk. |
Pack of 200 for $655.00
|
|
01-182-899
|
SiliCycle™
SPEP000101BB |
60 mg | 1 mL | 100 Pk. |
Pack of 100 for $278.00
|
|
01-182-900
|
SiliCycle™
SPEP000101BBL |
60 mg | 1 mL | 500 Pk. |
Pack of 500 for $1,320.00
|
|
01-182-905
|
SiliCycle™
SPEP000103BB |
60 mg | 3 mL | 50 Pk. |
Pack of 50 for $223.00
|
|
01-182-906
|
SiliCycle™
SPEP000103BBJ |
60 mg | 3 mL | 200 Pk. |
Pack of 200 for $823.00
|
|
01-182-912
|
SiliCycle™
SPEP000106BB |
60 mg | 6 mL | 30 Pk. |
Pack of 30 for $204.00
|
|
01-182-913
|
SiliCycle™
SPEP000106BBJ |
60 mg | 6 mL | 200 Pk. |
Pack of 200 for $1,012.00
|
|
01-182-901
|
SiliCycle™
SPEP000101C |
100 mg | 1 mL | 100 Pk. |
Pack of 100 for $321.00
|
|
01-182-907
|
SiliCycle™
SPEP000103C |
100 mg | 3 mL | 50 Pk. |
Pack of 50 for $231.00
|
|
01-182-908
|
SiliCycle™
SPEP000103CJ |
100 mg | 3 mL | 200 Pk. |
Pack of 200 for $876.00
|
|
01-182-909
|
SiliCycle™
SPEP000103CL |
100 mg | 3 mL | 500 Pk. |
Pack of 500 for $1,957.00
|
|
01-182-914
|
SiliCycle™
SPEP000106C |
100 mg | 6 mL | 30 Pk. |
Pack of 30 for $217.00
|
|
01-182-910
|
SiliCycle™
SPEP000103G |
200 mg | 3 mL | 50 Pk. |
Pack of 50 for $246.00
|
|
01-182-915
|
SiliCycle™
SPEP000106G |
200 mg | 6 mL | 30 Pk. |
Pack of 30 for $246.00
|
|
01-182-911
|
SiliCycle™
SPEP000103P |
500 mg | 3 mL | 50 Pk. |
Pack of 50 for $441.00
|
|
01-182-916
|
SiliCycle™
SPEP000106P |
500 mg | 6 mL | 30 Pk. |
Pack of 30 for $273.00
|
|
01-182-917
|
SiliCycle™
SPEP000106PJ |
500 mg | 6 mL | 200 Pk. |
Pack of 200 for $1,633.00
|
|
01-182-918
|
SiliCycle™
SPEP000112P |
500 mg | 12 mL | 20 Pk. |
Pack of 20 for $302.00
|
|
01-182-919
|
SiliCycle™
SPEP000112S |
1 g | 12 mL | 20 Pk. |
Pack of 20 for $329.00
|
|
01-182-921
|
SiliCycle™
SPEP000120S |
1 g | 25 mL | 20 Pk. |
Pack of 20 for $382.00
|
|
01-182-920
|
SiliCycle™
SPEP000112U |
2 g | 12 mL | 20 Pk. |
Pack of 20 for $671.00
|
|
01-182-922
|
SiliCycle™
SPEP000120U |
2 g | 25 mL | 20 Pk. |
Pack of 20 for $692.00
|
|
01-182-902
|
SiliCycle™
SPEP0001031A |
10 mg | 3 mL | 50 Pk. | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- For use with vacuum manifolds and automated SPE equipments
- Tightly packed and homogeneous silica bed for reliability and increased productivity
- Tight particle size distribution and very good packing with no fines, allowing excellent separation
- High recoveries and yields
- Reproducible flow rates lot-to-lot
- Flanged polypropylene tubes
- 20 μm polyethylene frits
Specifications
Polymer | |
Vacuum Manifolds or Automated SPE Equipment | |
SiliaPrepX™ | |
SPE Cartridges | |
Spherical |
Extraction of drugs and metabolites from biological fluids, API from tablets and cream | |
85 μm | |
DVB (Polystyrene-divinylbenzene) | |
Polypropylene (PP) tube, Polyethylene (PE) frits | |
60 Å |