Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Diagnostics™ Giemsa stain, modified
Used in staining blood or bone marrow films
$139.92
Specifications
Storage Requirements | 18-26°C |
---|---|
Color | Faint Blue to Dark Blue |
pH | 6.8 |
Synonym | Giemsa Solution |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
SDGS500
|
Sigma Diagnostics™
GS500500ML |
500mL |
Each of 1 for $139.92
|
|
|||||
Description
- When blood films are stained using Giemsa Stain, the nucleus and cytoplasm of white blood cells take on characteristic blue or pink coloration
- The use of purified eosin and thiazine dyes minimizes lot-to-lot variation
Specifications
18-26°C | |
Faint Blue to Dark Blue | |
6.8 | |
Giemsa Solution |
Safety and Handling
ShelfLife : Expiry date on the label.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title