Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Diagnostics™ Giemsa stain, modified
Used in staining blood or bone marrow films
Supplier: Sigma Diagnostics™ GS500500ML
Description
- When blood films are stained using Giemsa Stain, the nucleus and cytoplasm of white blood cells take on characteristic blue or pink coloration
- The use of purified eosin and thiazine dyes minimizes lot-to-lot variation
Specifications
500mL | |
Solution | |
For use in staining blood films or bone marrow films | |
6.8 |
18-26°C | |
0.4% (w/v) in buffered methanol solution, pH 6.8, with stabilizers | |
Faint Blue to Dark Blue | |
Giemsa Solution |
Safety and Handling
ShelfLife : Expiry date on the label.
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction