Learn More
MilliporeSigma™ Supelco™ SGE Syringes, Cone Tip
For use with Agilent/HP 7673, 7683 and 6850 ALS Autosamplers
Supplier: MilliporeSigma™ Supelco™ 21912
Description
- Designed and tested to meet critical autosampler specifications
- A unique plunger eliminates bending and seizing
- Needle point style withstands multiple and fast septum injections
- One of the most trouble free autosampler syringes available
Specifications
Stainless Steel (Tip) | |
SGE Syringe | |
1 | |
10 μL | |
Cone | |
Fixed |
26 G | |
SK-10 F-HP-0.47 | |
6 Pk. | |
42 mm | |
Agilent 7673, 7683, 6850 ALS Autosampler |