Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Set Hundreds Board
Two-sided.
$5.15
Alert:
Choking Hazard – small parts. Not for children under 3 years.
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S94303
|
Si Metric Manufacturing Limited
14520 |
Boards Only |
Pack of 10 for $5.15
|
|
|||||
S94301
|
Si Metric Manufacturing Limited
14500 |
Hundreds Board Set | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Plastic-coated cardboard
- Two-sided hundreds boards (12 × 11 in.)
- With 1 in. number squares
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title