Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Set Hundreds Board
Two-sided.
Supplier: Si Metric Manufacturing Limited 14520
Alert:
Choking Hazard – small parts. Not for children under 3 years.
Includes:
10 boards, 100 numbered and 25 blank plastic tiles, and a teacher's guide.
Description
- Plastic-coated cardboard
- Two-sided hundreds boards (12 × 11 in.)
- With 1 in. number squares
Specifications
Boards Only | |
Plastic | |
10/Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction