Learn More
Scott Safety™ E-Z Flo™ Regulators for Ska-Pak Supplied-Air Respirators
Designed for gloved hands, enables easy breathing, and has a two-step secure attachment.
$1534.23 - $1577.57
Specifications
Applications | Confined Space |
---|---|
For Use With (Equipment) | Ska-Pak Supplied Air Respirator |
Product Type | Regulator |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
SCT80422101
|
Scott Safety™
7100293240 |
Regulator only | N/A | N/A | |||||
Please contact your local Fisher Scientific sales representative for pricing | |||||||||
19-818-602
|
Scott Safety™
7100293244 |
Vibralert quick-disconnect |
Each for $1,577.57
|
|
|||||
SCT80437001
|
Scott Safety™
7012473991 |
Assembly |
Each for $1,534.23
|
|
|||||
Description
- Mask-mounted regulator maintains positive pressure during flows of up to 500 std. liters per minute
- Available in a continuous hose configuration, with an optional inline quick disconnect coupling
- The low-pressure hose equipped with a swivel attachment at the mask-mounted regulator
Specifications
Confined Space | |
Ska-Pak Supplied Air Respirator | |
Regulator |