Learn More
Scott Safety™ E-Z Flo™ Regulators for Ska-Pak Supplied-Air Respirators
Designed for gloved hands, enables easy breathing, and has a two-step secure attachment.
Supplier: Scott Safety™ 7100293244
Description
- Mask-mounted regulator maintains positive pressure during flows of up to 500 std. liters per minute
- Available in a continuous hose configuration, with an optional inline quick disconnect coupling
- The low-pressure hose equipped with a swivel attachment at the mask-mounted regulator
Specifications
Confined Space | |
Vibralert quick-disconnect | |
Regulator |
Ska-Pak Supplied Air Respirator | |
80437002 |