Learn More
Cole-Parmer™ RS-250 Series Reaction Station
Use the RS-250 Series Reaction Station to run multiple reactions in a single station, saving you time and money.
Supplier:  Cole-Parmer™ 3663012
 View more versions of this product
Catalog No. 13-245-156
Add to Cart
Description
Description
  • Excellent temperature uniformity across the block.
  • Soft-start feature minimizes splashing.
  • Run these reaction stations at the same temperature and stirring rates at several positions simultaneously.
  • They are great for a wide range of applications from simple synthesis to process optimization.
  • High-performance magnetic stirrers ensure maximum coupling between the stir bar in the sample and motor.
  • Easily set and view temperature and stir rate on the digital display.
  • Use the RS-232 or RS-485 connections to interface with a robotic workstation or a computer.
  • The well-insulated stations stay cool to the touch for safe handling.
  • PTFE coating on vessel block provides protection from chemical spills.
Specifications
Specifications
1 to 25 mL
4 kg
302°F
115 V
400 to 2000 rpm
150°C
10 x 1 to 25 mL Tubes
Reaction Station
12 x 3 1/8 x 5 7/8 in.
8.8 lb.
Product Suggestions
Product Suggestions