Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotix™ Robotic aTIP Filter Pipette Tips
For use with Agilent™ Velocity 11™ VPrep and Bravo, 384-Head platforms.
$681.50 - $789.50
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Color | Clear |
For Use With (Equipment) | 96/384-well Plates |
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
12-111-059
|
Biotix™
VB00303SC |
30 μL |
Pack of 10 for $681.50
Case of 5 PK for $3,410.00 |
|
12-111-060
|
Biotix™
VB00303FC |
15 μL |
Pack of 10 for $789.50
Case of 5 PK for $3,950.00 |
|
Description
- Made from clear polypropylene
- Compatible with 96-and 384-well plates
Specifications
Filtered | |
Sterile | |
Clear | |
96/384-well Plates |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title