Learn More
Biotix™ Robotic aTIP Filter Pipette Tips
For use with Agilent™ Velocity 11™ VPrep and Bravo, 384-Head platforms.
Supplier:  Biotix™ VB00303FC
 View more versions of this product
Catalog No. 12-111-060
Add to cart
Description
Description
  • Made from clear polypropylene
  • Compatible with 96-and 384-well plates
Specifications
Specifications
15 μL
Sterile
96/384-well Plates
Filtered
Clear
Polypropylene