Learn More
Tocris™ Rhein
FTO (mRNA N6-methyladenine demethylase) inhibitor; also inhibits ALKBH2 and ALKBH3
Supplier:  Tocris™ 6867/50
 View more versions of this product
Catalog No. 68-675-0
Add to Cart
This item is not returnable. View return policy
Description
Description
Rhein is a FTO (mRNA N6-methyladenine demethylase) inhibitor (IC50 = 1.2 - 21 μM; Ki = 1.8 μM). Competitively binds enzyme at active site preventing binding of N6-methyladenosine (m6A) substrate and resulting in accumulation of DNA damage in cells. Also inhibits ALKBH3 and ALKBH2 (IC50 values are 5.3 and 9.1 μM, respectively). Sensitizes glioblastoma cells to alkylating agents. Active in vivo.
Specifications
Specifications
50 mg
284.22
98%
C15H8O6
Inhibitor
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C