Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BrandTech™ Replacement Seals, Pump Assemblies and Valve Block Caps for BRAND seripettor and seripettor pro Bottletop Dispensers
Accessories for BRAND seripettor and seripettor pro Bottletop Dispensers
Supplier: BrandTech™ 704549
Description
- 6788 - EPDM seals for seripettor dispensers, pack of 5
- 704536 - autoclavable filling tube for seripettor, 250mm, each
- 704538 - autoclavable filling tube for seripettor, 500mm, each
- 704542 - pump assembly for 1-10mL seripettor
- 704544 - pump assembly for 2.5-25mL seriipettor
- 704549 - Pump assembly for 2.5-25mL seripettor pro
- 704552 - seripettor valve block cap for autoclaving. Fits 1-10mL seripettor
- 704554 seripettor valve block cap for autoclaving. Fits 2.5-10mL seripettor
Specifications
Pump Assembly | |
Dark Blue, Red, Translucent Amber | |
Seripettor™ pro, 25 mL |
Not Autoclavable | |
Dispensing liquid | |
Pump assembly only |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction