Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BrandTech™ Replacement Seals, Pump Assemblies and Valve Block Caps for BRAND seripettor and seripettor pro Bottletop Dispensers
Accessories for BRAND seripettor and seripettor pro Bottletop Dispensers
$19.50 - $360.00
Catalog Number | Mfr. No. | Color | Product Type | Autoclavable | Includes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Product Type | Autoclavable | Includes | Price | Quantity & Availability | ||
13-880-707
|
BrandTech™
6788 |
Black | Sealing Washer | Autoclavable | Five sealing washers |
Pack of 5 for $19.50
|
|
||
13-880-731
|
BrandTech™
704544 |
Clear, Blue, Red | Pump Assembly | Not Autoclavable | Pump assembly only |
Each for $235.00
|
|
||
13-880-732
|
BrandTech™
704549 |
Dark Blue, Red, Translucent Amber | Pump Assembly | Not Autoclavable | Pump assembly only |
Each for $360.00
|
|
||
Description
- 6788 - EPDM seals for seripettor dispensers, pack of 5
- 704536 - autoclavable filling tube for seripettor, 250mm, each
- 704538 - autoclavable filling tube for seripettor, 500mm, each
- 704542 - pump assembly for 1-10mL seripettor
- 704544 - pump assembly for 2.5-25mL seriipettor
- 704549 - Pump assembly for 2.5-25mL seripettor pro
- 704552 - seripettor valve block cap for autoclaving. Fits 1-10mL seripettor
- 704554 seripettor valve block cap for autoclaving. Fits 2.5-10mL seripettor
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title