Learn More
Thermo Scientific™ Remel™ BactiSwab™ Wire Shaft Amies Clear
Supplied with a mini-tip swab suitable for ear, nose, throat, eye, male urethral and pediatric specimen collection.
$240.00 - $411.00
Specifications
Format | Tube |
---|---|
Media Type | Amies Clear |
Closure Color | Orange |
Description | Amies Clear |
Quantity | 1 Tube/Envelope |
Description
Thermo Scientific™ Remel BactiSwab Gel: Wire Shaft Amies Clear is supplied with a mini-tip swab suitable for ear, nose, throat, eye, male urethral and pediatric specimen collection.
Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing gel transport medium with or without charcoal.- Straight aluminum shaft
- Orange cap
- Round-bottom tube for easy racking and seamless molding to ensure strength and integrity
- Exclusive zip-seal packaging to promote optimum media performance
- Color-coded cap with protective overskirt
- Inert, non-toxic rayon swab tip for good sample retrieval and absorption
Specifications
Tube | |
Orange | |
1 Tube/Envelope |
Amies Clear | |
Amies Clear | |
Wire |