Learn More
Thermo Scientific™ Remel™ BactiSwab™ Wire Shaft Amies Clear
Supplied with a mini-tip swab suitable for ear, nose, throat, eye, male urethral and pediatric specimen collection.
Supplier:  Thermo Scientific™ R723020
 View more versions of this product
Catalog No. R723020
Add to Cart
This item is not returnable. View return policy
Description
Description
Thermo Scientific™ Remel BactiSwab Gel: Wire Shaft Amies Clear is supplied with a mini-tip swab suitable for ear, nose, throat, eye, male urethral and pediatric specimen collection.
Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing gel transport medium with or without charcoal.
  • Straight aluminum shaft
  • Orange cap
  • Round-bottom tube for easy racking and seamless molding to ensure strength and integrity
  • Exclusive zip-seal packaging to promote optimum media performance
  • Color-coded cap with protective overskirt
  • Inert, non-toxic rayon swab tip for good sample retrieval and absorption
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tube
Orange
100/Pk.
Wire
Amies Clear
Amies Clear
1 Tube/Envelope