Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Z-Leu-Leu-Leu-B(OH)2 (MG-262)
Highly potent and selective cell-permeable inhibitor of the proteasome
Specifications
Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
---|---|
Molecular Weight (g/mol) | 491.44 |
Inhibitors | Proteasome |
Description
- This peptide boronic acid is structurally similar to the peptide aldehyde MG-132 and also inhibits the chymotrypsin-like peptidase activity of the proteasome, but with much higher potency
- These inhibitors are reversible and also inhibit the activity of calpains and cathepsins
- Purity >95% by HPLC
- Structure confirmed by 1H-NMR
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
491.44 | |
Proteasome |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title